missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific Chemicals Hydroxy Naphthol Blue, Disodium Salt, Pure

Supplier:  Thermo Scientific Chemicals 204880050

 View more versions of this product

Catalog No. AC204880050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Hydroxy Naphthol Blue, disodium salt
165660-27-5
MFCD00004075
Solubility in water: soluble.
Authentic
3-hydroxy-4-[(2Z)-2-(2-oxo-4-sulfonaphthalen-1-ylidene)hydrazinyl]naphthalene-2,7-disulfonic acid;sodium
131849366
ACS Reagent
(for calcium determination) passes
5 g
274.0°C
C20H12N2Na2O11S3
2,2'-Dihydroxy-1,1'-azonaphthalene-3',4,6'-trisulfonic acid disodium salt
LXJHFXHZWYBBKJ-LUUCHEBKSA-N
C1=CC=C2C(=C1)C(=CC(=O)C2=NNC3=C4C=CC(=CC4=CC(=C3O)S(=O)(=O)O)S(=O)(=O)O)S(=O)(=O)O.[Na].[Na]
600.495
598.47
Glass bottle
Blue to Purple
Crystalline Powder or Crystals

Chemical Identifiers

165660-27-5
600.495
LXJHFXHZWYBBKJ-LUUCHEBKSA-N
131849366
C1=CC=C2C(=C1)C(=CC(=O)C2=NNC3=C4C=CC(=CC4=CC(=C3O)S(=O)(=O)O)S(=O)(=O)O)S(=O)(=O)O.[Na].[Na]
C20H12N2Na2O11S3
MFCD00004075
2,2'-Dihydroxy-1,1'-azonaphthalene-3',4,6'-trisulfonic acid disodium salt
3-hydroxy-4-[(2Z)-2-(2-oxo-4-sulfonaphthalen-1-ylidene)hydrazinyl]naphthalene-2,7-disulfonic acid;sodium
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Room tempature

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only