missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

[Hydroxy(tosyloxy)iodo]benzene, 97+%, Thermo Scientific Chemicals

$89.11 - $89.11

Chemical Identifiers

CAS 27126-76-7
Molecular Formula C13H13IO4S
Molecular Weight (g/mol) 392.21
MDL Number MFCD00011547
InChI Key LRIUKPUCKCECPT-UHFFFAOYSA-N
Synonym hydroxy tosyloxy iodo benzene, htib, koser's reagent, phenyliodosohydroxy tosylate, koser reagent, unii-2eoa97b14a, hydroxy p-tolylsulfonyl oxy iodo benzene, hydroxy 4-methylbenzenesulfonato-o phenyliodine, iodosobenzene-i-mono-p-toluenesulfonate, hydroxy phenyl 4-methylbenzenesulfonoperoxoyl iodide
PubChem CID 325434
IUPAC Name [hydroxy(phenyl)-$l^{3}-iodanyl] 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OI(C2=CC=CC=C2)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC389610050
SDS View Documents
Thermo Scientific Chemicals
389610050
5 g Glass bottle
Each for $89.11
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

oxidation reagent
Specifications

Chemical Identifiers

27126-76-7
392.21
LRIUKPUCKCECPT-UHFFFAOYSA-N
325434
CC1=CC=C(C=C1)S(=O)(=O)OI(C2=CC=CC=C2)O
C13H13IO4S
MFCD00011547
hydroxy tosyloxy iodo benzene, htib, koser's reagent, phenyliodosohydroxy tosylate, koser reagent, unii-2eoa97b14a, hydroxy p-tolylsulfonyl oxy iodo benzene, hydroxy 4-methylbenzenesulfonato-o phenyliodine, iodosobenzene-i-mono-p-toluenesulfonate, hydroxy phenyl 4-methylbenzenesulfonoperoxoyl iodide
[hydroxy(phenyl)-$l^{3}-iodanyl] 4-methylbenzenesulfonate

Specifications

27126-76-7
100.0
Yellow
97+%
C13H13IO4S
5 g
hydroxy tosyloxy iodo benzene, htib, koser's reagent, phenyliodosohydroxy tosylate, koser reagent, unii-2eoa97b14a, hydroxy p-tolylsulfonyl oxy iodo benzene, hydroxy 4-methylbenzenesulfonato-o phenyliodine, iodosobenzene-i-mono-p-toluenesulfonate, hydroxy phenyl 4-methylbenzenesulfonoperoxoyl iodide
CC1=CC=C(C=C1)S(=O)(=O)OI(C2=CC=CC=C2)O
392.21
392.21
Crystalline Powder
97.0
131.0°C to 137.0°C
Authentic
Glass bottle
MFCD00011547
15, 5369
LRIUKPUCKCECPT-UHFFFAOYSA-N
[hydroxy(phenyl)-$l^{3}-iodanyl] 4-methylbenzenesulfonate
325434
97+%
[Hydroxy(tosyloxy)iodo]benzene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only