missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydroxylamine Hydrochloride (Suitable for Mercury Determination/Certified ACS), Fisher Chemical™

Supplier:  Thermo Fisher Scientific H3301

 View more versions of this product

Catalog No. H330-1



Explore available promotions

Description

Description

Specifications

Specifications

Hydroxylamine Hydrochloride
Suitable for Mercury Determination
White
Solid
≥96 %
NH2OH·HCl
2923
WTDHULULXKLSOZ-UHFFFAOYSA-N
hydroxylamine hydrochloride
443297
69.49
Certified ACS
Poly Bottle
1.6700g/cm³
155°C
5470-11-1
2.5 to 3.5
1 kg
ClH4NO
MFCD00051089
hydroxylamine hydrochloride, hydroxylamine hcl, hydroxylammonium chloride, oxammonium hydrochloride, hydroxylamine, hydrochloride, hydroxyamine hydrochloride, hydroxylamine chloride 1:1, hydroxylamine, hydrochloride 1:1, hydroxyamine hcl, oxammonium hci
Cl.NO
69.49
CHEBI:5807
≥96%
Pass Test
Negligible

Chemical Identifiers

5470-11-1
69.49
WTDHULULXKLSOZ-UHFFFAOYSA-N
443297
hydroxylamine hydrochloride
ClH4NO
MFCD00051089
hydroxylamine hydrochloride, hydroxylamine hcl, hydroxylammonium chloride, oxammonium hydrochloride, hydroxylamine, hydrochloride, hydroxyamine hydrochloride, hydroxylamine chloride 1:1, hydroxylamine, hydrochloride 1:1, hydroxyamine hcl, oxammonium hci
CHEBI:5807
Cl.NO
Videos
Safety and Handling

Safety and Handling

DOTInformation : DOT Class 8, : Corrosive

SDS Documents

Documents

Promotions

Promotions