missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hydroxylamine Hydrochloride (Suitable for Mercury Determination/Certified ACS), Fisher Chemical™
Supplier: Thermo Fisher Scientific H330500
Specifications
Hydroxylamine Hydrochloride | |
Suitable for Mercury Determination | |
White | |
Solid | |
≥96 % | |
NH2OH·HCl | |
2923 | |
WTDHULULXKLSOZ-UHFFFAOYSA-N | |
hydroxylamine hydrochloride | |
443297 | |
69.49 | |
Certified ACS | |
Poly Bottle | |
1.6700g/cm³ |
155°C | |
5470-11-1 | |
2.5 to 3.5 | |
500 g | |
ClH4NO | |
MFCD00051089 | |
hydroxylamine hydrochloride, hydroxylamine hcl, hydroxylammonium chloride, oxammonium hydrochloride, hydroxylamine, hydrochloride, hydroxyamine hydrochloride, hydroxylamine chloride 1:1, hydroxylamine, hydrochloride 1:1, hydroxyamine hcl, oxammonium hci | |
Cl.NO | |
69.49 | |
CHEBI:5807 | |
≥96% | |
Pass Test | |
Negligible |
Chemical Identifiers
5470-11-1 | |
69.49 | |
WTDHULULXKLSOZ-UHFFFAOYSA-N | |
443297 | |
hydroxylamine hydrochloride |
ClH4NO | |
MFCD00051089 | |
hydroxylamine hydrochloride, hydroxylamine hcl, hydroxylammonium chloride, oxammonium hydrochloride, hydroxylamine, hydrochloride, hydroxyamine hydrochloride, hydroxylamine chloride 1:1, hydroxylamine, hydrochloride 1:1, hydroxyamine hcl, oxammonium hci | |
CHEBI:5807 | |
Cl.NO |
Safety and Handling
DOTInformation : DOT Class 8, : Corrosive