missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydroxynaphthol Blue (Certified), Fisher Chemical

Used as an indicator for EDTA titrimetric determination of calcium. Color changes from reddish pink to blue

Supplier:  Thermo Fisher Scientific H346100

Catalog No. H346-100



Explore available promotions

Description

Description

Specifications

Specifications

Hydroxynaphthol Blue
C20H11N2Na3O11S3
1-(2-Hydroxy-4-sulfo-1-naphthylazo)-2-naphthol-3,6-disulfonic acid trisodium salt
JQECMNGWLIFQLL-UHFFFAOYSA-K
trisodium;3-hydroxy-4-[2-(2-oxo-4-sulfonatonaphthalen-1-ylidene)hydrazinyl]naphthalene-2,7-disulfonate
44134769
Certified
Pass Test
6, 7
Solid
63451-35-4
MFCD00036393
Pass Test
C1=CC=C2C(=C1)C(=CC(=O)C2=NNC3=C4C=CC(=CC4=CC(=C3O)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+].[Na+]
620.46
620.47
Glass Bottle
Gray
100 g

Chemical Identifiers

63451-35-4
620.46
JQECMNGWLIFQLL-UHFFFAOYSA-K
44134769
C1=CC=C2C(=C1)C(=CC(=O)C2=NNC3=C4C=CC(=CC4=CC(=C3O)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+].[Na+]
C20H11N2Na3O11S3
MFCD00036393
1-(2-Hydroxy-4-sulfo-1-naphthylazo)-2-naphthol-3,6-disulfonic acid trisodium salt
trisodium;3-hydroxy-4-[2-(2-oxo-4-sulfonatonaphthalen-1-ylidene)hydrazinyl]naphthalene-2,7-disulfonate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

SDS Documents

Documents

Promotions

Promotions