missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydroxyurea, 98%, Thermo Scientific Chemicals

Hydroxyurea, 98%, CAS # 127-07-1, is a compound with antineoplastic, immunomodulatory, radical scavenger, and antimitotic activities. It shows inhibitory activity against nucleic acid synthesis.

$218.49 - $218.49

Chemical Identifiers

CAS 127-07-1
Molecular Formula CH4N2O2
Molecular Weight (g/mol) 76.04
MDL Number MFCD00007943
InChI Key VSNHCAURESNICA-UHFFFAOYSA-N
Synonym hydroxycarbamide, n-hydroxyurea, hydrea, 1-hydroxyurea, oxyurea, carbamoyl oxime, biosupressin, hydroxycarbamine, onco-carbide, carbamohydroxamic acid
PubChem CID 3657
ChEBI CHEBI:44423
IUPAC Name hydroxyurea
SMILES C(=O)(N)NO
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC151680050
SDS View Documents
Thermo Scientific Chemicals
151680050
5 g Glass bottle
Each for $218.49
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Hydroxyurea is a member of the class of urea that shows antineoplastic, immunomodulatory, radical scavenger, and antimitotic activities.

• It has inhibitory activity against nucleic acid synthesis.

• It also inhibits ribonucleoside diphosphate reductase, an enzyme necessary to convert ribonucleoside diphosphate into deoxyribonucleoside diphosphate, preventing cells from leaving the G1/S phase of the cell cycle

Applications

• Hydroxyurea has an anti-retroviral activity, inhibiting human immunodeficiency virus type 1 (HIV-1) DNA synthesis in peripheral blood lymphocytes, decreasing the amount of intracellular deoxynucleotides

• This compound also has a radio sensitizing activity, maintaining cells in the radiation-sensitive G1 cell cycle phase and, consequently, interfering with DNA repair

• It has been used for cell cycle synchronization in human breast carcinoma cells

• This compound presents inhibitory activity in cell adhesion and motility assay in ccRCC Caki (HTB-47) and CD9 knockdown cells

Specifications

Chemical Identifiers

127-07-1
76.04
VSNHCAURESNICA-UHFFFAOYSA-N
3657
hydroxyurea
CH4N2O2
MFCD00007943
hydroxycarbamide, n-hydroxyurea, hydrea, 1-hydroxyurea, oxyurea, carbamoyl oxime, biosupressin, hydroxycarbamine, onco-carbide, carbamohydroxamic acid
CHEBI:44423
C(=O)(N)NO

Specifications

127-07-1
White
Authentic
Glass bottle
HONHCONH2
5 g
15, 4888
Solubility in water: freely soluble. Other solubilities: freely soluble in hot alcohol
C(=O)(N)NO
76.04
CHEBI:44423
98%
Hydroxyurea
142°C to 146°C
1% max. (1 g, 60°C) (< 0.7 kPa)
97.5% min. (on dried substance) (Iodometry)
CH4N2O2
MFCD00007943
03, 95
hydroxycarbamide, n-hydroxyurea, hydrea, 1-hydroxyurea, oxyurea, carbamoyl oxime, biosupressin, hydroxycarbamine, onco-carbide, carbamohydroxamic acid
VSNHCAURESNICA-UHFFFAOYSA-N
hydroxyurea
3657
76.04
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause genetic defects.
May damage the unborn child.

GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 204-821-7

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only