Ansell HyFlex™ 11-250 Cut-Resistant Protective Arm Sleeve without Thumbslot
Gray arm protection sleeves that combine cut protection with unrivaled comfort.
Supplier:  Ansell 11250180WVEND
 View more versions of this product
Catalog No. 17004288
Explore available promotions
Description
Description
  • INTERCEPT™ Seamless High Performance Polyethylene (HPPE)
  • Seamless Design
  • Silicone-Free
  • Red and black over edge is to indicate wide (black) and narrow (red) for easy laundry sorting purpose
Specifications
Specifications
Knit
Wide Sleeve
HPPE Yarn
18 in.