Ansell Hyflex™ 11-561 Vend Pack Gloves
Our lightest EN ISO Cut C and ANSI/ISEA 105-2024 Cut A3 glove. Offering 2 times greater cut resistance.
$133.10 - $133.10
Specifications
Certifications/Compliance EN 388:2016, Dermatologically Tested, Ecovadis label, EN 388:2016 +A1:2018, EN ISO 21420:2020, Food Contact, OEKO-TEX™ Centexbel, Category II
Construction Knit
Cuff Material Nylon, HPPE, Basalt, Spandex, Polyester
Coating Material Nitrile
Color Blue
View More Specs
Products 8
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
17004253
View Documents
Ansell
11561VP050
5
Pack of 12 Each for $133.10
Case of 12 Pack for $1,526.76
17004254
View Documents
Ansell
11561VP060
6
Pack of 12 Each for $133.10
Case of 12 Pack for $1,526.76
17004255
View Documents
Ansell
11561VP070
7
Pack of 12 Each for $133.10
Case of 12 Pack for $1,526.76
17004256
View Documents
Ansell
11561VP080
8
Pack of 12 Each for $133.10
Case of 12 Pack for $1,526.76
17004257
View Documents
Ansell
11561VP090
9
Pack of 12 Each for $133.10
Case of 12 Pack for $1,526.76
17004258
View Documents
Ansell
11561VP100
10
Pack of 12 Each for $133.10
Case of 12 Pack for $1,526.76
17004259
View Documents
Ansell
11561VP110
11
Pack of 12 Each for $133.10
Case of 12 Pack for $1,526.76
17004260
View Documents
Ansell
11561VP120
12
Pack of 12 Each for $133.10
Case of 12 Pack for $1,526.76
Description
Description
  • Ultra-lightweight design: 20% lighter for unparalleled comfort and dexterity
  • EN CUT ISO C and ANSI/ ISEA 105-2024 CUT A3 protection: 2x greater cut resistance
  • Improved FORTIX™ Technology: up to 20% greater durability
Specifications
Specifications
EN 388:2016, Dermatologically Tested, Ecovadis label, EN 388:2016 +A1:2018, EN ISO 21420:2020, Food Contact, OEKO-TEX™ Centexbel, Category II
Nylon, HPPE, Basalt, Spandex, Polyester
Blue
Specific
Nylon, HPPE, Spandex, Polyester, Nitrile
4 x 42 C
Unisex
Knit
11-561
Reinforced
Knit
Nitrile
ANSI A3
No
Cut-resistant Gloves
Handling metal sheets and panels, Handling parts with sharp, rough edges, Final assembly, Handling materials with sharp or rough edges, Assembly and inspection of components, Applying finish to materials, products, Compatible with consumer and industrial touchscreens
Blue
Nylon, HPPE, Basalt, Spandex, Polyester
ANSI/ISEA 105-16 Level 4
15
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'