missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ansell HyFlex™ 11-751 Medium Duty Abrasion-Resistant Gloves, INTERCEPT™ Technology
Level C cut protection gloves offer enhanced abrasion resistance for extended wear.
Supplier: Ansell 11751090
Description
- INTERCEPT Technology™ for high level of cut protection
- High durability PU coating, increased levels of abrasion resistance
- Dirt masking design allows for extended product use
- In compliance with the standards EN420 (General requirements) and EN388 (Mechanical hazards)
- Also available in vendor pack
Specifications
ANSI 6 | |
Knit | |
Black | |
Specific | |
Cut-resistant Gloves | |
EN 388:4X43C | |
Knit | |
11751090 | |
HyFlex™ 11-751 | |
22.4 cm |
CE, EN 420:2003 + A1:2009; Category II | |
Polyurethane | |
ANSI A4 | |
Yes | |
9 | |
General maintenance, hardware assembly, machining, stamping, cutting, raw material handling | |
HPPE, Fiberglass, Nylon, Spandex | |
12 Pairs/Bag, 12 Bags/Cs. | |
8.8 in. |