Ansell HyFlex™ 11-757 Cut-Resistant Touchscreen Gloves
Ultralight ANSI A7 / ISO F-level cut-resistant touchscreen glove.
$176.63 - $176.63
Specifications
Coating Material | Polyurethane |
---|---|
Color | Black |
Cut Resistance Rating | ANSI A7 |
Latex Free | Yes |
Material | Nylon, Stainless Steel Fiber, Polyester, Tungsten |
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
17004275
|
Ansell
11757060VP |
6 |
Pack of 12 Each for $176.63
Case of 12 Pack for $2,026.08
|
|
|||||
17004276
|
Ansell
11757070VP |
7 |
Pack of 12 Each for $176.63
Case of 12 Pack for $2,026.08
|
|
|||||
17004277
|
Ansell
11757080VP |
8 |
Pack of 12 Each for $176.63
Case of 12 Pack for $2,026.08
|
|
|||||
17004278
|
Ansell
11757090VP |
9 |
Pack of 12 Each for $176.63
Case of 12 Pack for $2,026.08
|
|
|||||
17004279
|
Ansell
11757100VP |
10 |
Pack of 12 Each for $176.63
Case of 12 Pack for $2,026.08
|
|
|||||
17004280
|
Ansell
11757110VP |
11 |
Pack of 12 Each for $176.63
Case of 12 Pack for $2,026.08
|
|
|||||
Description
- Extreme cut protection featuring INTERCEPT™ Technology while maintaining enhanced flexibility and tactility
- Touchscreen compatible to ensure that workers do not need to change or remove gloves between tasks
- High dexterity combined with unmatched comfort and precision while achieving excellent abrasion resistance
- Tailored fit at the base of the pinky finger better matches natural hand contours
- Dermatologically approved by Dermatest™
Specifications
Polyurethane | |
ANSI A7 | |
Nylon, Stainless Steel Fiber, Polyester, Tungsten |
Black | |
Yes |