Hygiena Aquasnap™ ATP Water Testing Device
User-friendly, self-contained ATP water sampling device used with the Hygiena luminometers
Supplier:  Hygiena AQ100X
Catalog No. 14-037-768
Explore available promotions
Requires : Luminometer
Description
Description
AquaSnap is easy to use, economical, and gives real time results. The specifically designed dipper tip collects 100μl of water ensuring consistent sample collection.
AquaSnap Total measures both free ATP in solution and microbial ATP. AquaSnap Total contains a detergent to release ATP that is bound to microbial or organic matter and inside microbes. AquaSnap Free measures dissolved ATP that is free in solution. It is intended for use in food and beverage processing to detect organic contamination in rinse water samples from CIP systems, tanks and pipes. The difference between Total and Free ATP provides a measure of microbial contamination in the sample.
  • Patented Snap-Valve™ technology and self-contained liquid stable reagent minimizes costs and maximizes performance
  • Write-on swab label
  • Results in 15 seconds
  • Unprecedented accuracy and reproducibility
  • Honeycomb-shaped collection tip collects 100μL of sample - no pipetting
  • Compatible with SystemSURE Plus and EnSURE luminometers