Hygiena Q-Swab™
All-in-one dilution and delivery device system for environmental sample collection
Supplier:  Hygiena QS1200
 View more versions of this product
Catalog No. 14-037-764
Explore available promotions
Description
Description
Once the sample area has been swabbed, snapping the Snap-Valve releases 1mL of broth into swab shaft, neutralizing residual sanitizers and facilitating the recovery of bacteria. The sample is then ready to be poured onto dehydrated media or streaked onto agar plates for culturing.
  • Each broth is gamma irradiated and aseptically filled into a self-contained reservoir at the tip of the device
  • Ideal for surface sampling in the food and beverage industry
  • Eliminate the need for pipettes and broth preparation
  • Swabs can be used either wet or dry
  • 1mL sample perfectly hydrates dried media
  • Neutralizes residual sanitizers remaining on surface
  • Environmental microbial sample collection and transport
  • All-in-one sample collection system
  • 250 devices per case
Specifications
Specifications
Ideal for surface sampling in the food and beverage industry
2°C to 8°C
Letheen Broth