Remco™ Hygienic Hi-Flex Wall Bracket System
This hygienic, modular wall bracket provides a flexible solution for storing your cleaning tools and utensils in any area of your site. The rail easily attaches to four wall mounts by sliding it on from the right or left side.
Supplier:  Remco™ REM10114
 View more versions of this product
Catalog No. 17228003
Explore available promotions
Description
Description
  • Grip band modules and hook modules can be clicked on the rail and locked in place using one of many small nothces in the rail
  • Up to ten hanging modules can be placed on the rail (or up to five grip band modules)
  • System comes with three grip band modules and two hooks
  • Easy to dissemble for cleaning and replacement
Specifications
Specifications
Wall Bracket
Polypropylene
16.5 in.
3.4 in.
Red
1.9 in.
0.63 lb.