missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Remco™ Hygienic Wall Bracket, Double Hook Module
This double hook module is designed for hanging 1-2 cleaning tools that have a hanging hole, or products with a D- or T- shaped handle.
Supplier: Remco™ 10148
Description
- Hooks are slid onto the supplied double-base/spacer from either side
- Hook module can hold products weighing up to 3 kg
- Easy to dissemble for cleaning or replacement
Specifications
Wall Bracket | |
Polypropylene | |
3.2 in. | |
3.1 in. |
Purple | |
1.9 in. | |
0.12 lb. |