Epredia™ Hyperclean™ TruAir™ Ductless Fume Hood Accessories
Add convenience and functionality to your fume hood with Epredia™ Hyperclean™ TruAir™ Ductless Fume Hood Accessories.
Supplier:  Epredia™ B1011481
 View more versions of this product
Catalog No. B1011481
Explore available promotions
Description
Description
Accessory options include drip trays, base stands, air flow indicator, and filters.
Specifications
Specifications
Polypropylene
Drip Tray 48 Polypropylene
B1115481, B1115483, B1220481, B1220483