missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ HyPerforma™ 50 to 500L Tank Liners, 100L
Thermo Scientific™ HyPerforma™ 50 to 500L Tank Liners are available with or without bottom drain and are provided sterile for handling fluids in bioproduction applications.
Supplier: Thermo Scientific™ SH3064705
Description
Standard Thermo Scientific tank liners feature an open top for maximum recovery, with drum dolly available to provide mobility.
Features:
- Minimizes bioburden due to construction in an ISO 7 cleanroom under sterile cGMP conditions.
- Eliminates post-use cleaning steps required with re-useable containers and reduces cross-contamination risks.
Applications of the Thermo Scientific tank liner:
- Removes the need for tank cleaning and helps reduce cycle times.
- Facilitates hydration of powdered media and buffers, and pooling of non-sterile solutions and fluids.
Specifications
100 L | |
Packed and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area. | |
Irradiated | |
CX3-9 |
Packages are placed in strong cardboard cartons with product and lot identification. | |
SV30112 | |
HyPerforma™ |