missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ HyPerforma™ 50 to 500L Tank Liners, 50L

Thermo Scientific™ HyPerforma™ 50 to 500L Tank Liners are available with or without bottom drain and are provided sterile for handling fluids in bioproduction applications.

Supplier:  Thermo Scientific™ SH3064704

Catalog No. SH3064704

Explore available promotions

Description

Description

Thermo Scientific™ HyPerforma™ 50 to 500L Tank Liners are available with or without bottom drain and are provided sterile for handling fluids in bioproduction applications.

Tank Liner Key Features:

  • Available in sizes that fit into industry standard cylindrical tanks and in Thermo Scientific HyPerforma Drums (SV3011X, SV50076, and SV50517 series)
  • Gusset design allows for a close fit to the tank
  • Chamber constructed of Thermo Scientific CX3-9 film, which provides the same contact surface as other Thermo Scientific BPCs
  • Supplied gamma irradiated
  • Customization options are available by contacting one of our Thermo Scientific Representatives today

Standard Tank Liners for your liquid applications available in-stock in the following configurations:

SH30647.01 to .03:

  • 50 to 200L Tank Liner
  • Support Container Needed: Design to fit into HyPerforma Flat Bottom PE Drums (SV50076 series)
  • Quantity: Each = 1 Liner

SH30647.04 to .07:

  • 50 to 500L Tank Liner
  • Support Container Needed: Designed to fit into Industry Standard Cylindrical Tanks (SV3011X series)
  • Quantity: Each = 1 Liner

SH30646 Series:

  • 50 to 200L Tank Liner with One Bottom Drain
  • Bottom Lineset 1: 72in. (182.9cm) of tubing leading to a Quick Disconnect (MPX series) Insert (male) connector
  • Support Container Needed: Designed to fit into HyPerforma Conical Bottom PE Drums (SV50517 series)

Ordering Information:

Supplied double polybagged and irradiated with Certificate of Analysis.

Recommended for:

  • Hydrations of powdered media and buffers
  • Pooling of non-sterile solutions and fluids
  • Non-aseptic collection of waste liquids from separation and purification systems

To order, use the part numbers listed in the above table.

Specifications

Specifications

50 L
Tank Liner
Packed and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area.
Irradiated
Fewer resources used and less waste
Sterile
Plastic
Packages are placed in strong cardboard cartons with product and lot identification.
SV30111
HyPerforma™
CX3-9
Videos
SDS
Documents

Documents

Promotions

Promotions