missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hypericin, 98%, Thermo Scientific Chemicals

Hypericin, CAS # 548-04-9, is an anthraquinone derivative that shows antidepressant, antiviral, antineoplastic, and immunostimulant activities.

Supplier:  Thermo Scientific Chemicals H26425MC

 View more versions of this product

Catalog No. AAH26425MC


Explore available promotions

Description

Description

Inhibitor of protein kinase C; induces photosynthesized apoptosisHypercin acts as an antibiotic and antiviral. It is involved in the inhibition of enzyme dopamine beta-hydroxylase action. It also exhibits an anti-HIV and anticancer drug. Further, it is useful in photodynamic therapy. In addition to this, it serves as an inhibitor of protein kinase C which induces photosynthesized apoptosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Hypericin is an anthraquinone derivative found in the yellow flower of Hypericum perforatum (also known as St. John's wort)
  • This compound has the ability to inhibit protein kinase C, inducing photosynthesized apoptosis

Application

  • It shows antibiotic and antiviral activities
  • The antidepressant activity of this compound could be related to its ability to inhibit the neuronal uptake of serotonin, norepinephrine, dopamine, gamma-aminobutyric acid (GABA), and L-glutamate
  • Since this compound is able to inhibit the assembly and shedding of virus particles in infected cells, hypericin may also prevent the replication of encapsulated viruses
  • It has been described that this compound can also exhibit anti-HIV and anticancer activities
  • Hypericin also displays an effective phototoxic activity and is able to trigger apoptotic pathways, resulting in the formation of reactive oxygen species
Specifications

Specifications

Hypericin
∼320°C (decomposition)
MFCD00016683
1917913
14,4863
Soluble in ethanol,dimethyl sulfoxide,methanol,acetone,ethylmethylketone,pyridine and sodium hydroxide. Insoluble in water.
CC1=CC(=C2C3=C1C4=C5C(=C(C=C4C)O)C(=O)C6=C(C=C(C7=C6C5=C3C8=C7C(=CC(=C8C2=O)O)O)O)O)O
5281051
504.44
548-04-9
C30H16O8
100 mg
Light sensitive
hypericin, hypericum red, hipericina, hypericine, hyperizin, cyclosan, 1,3,4,6,8,13-hexahydroxy-10,11-dimethylphenanthro 1,10,9,8-opqra perylene-7,14-dione, cyclo-werol, hypericum extract, vimrxyn
BTXNYTINYBABQR-UHFFFAOYSA-N
504.45
CHEBI:5835
98%

Chemical Identifiers

548-04-9
504.45
BTXNYTINYBABQR-UHFFFAOYSA-N
5281051
CC1=CC(=C2C3=C1C4=C5C(=C(C=C4C)O)C(=O)C6=C(C=C(C7=C6C5=C3C8=C7C(=CC(=C8C2=O)O)O)O)O)O
C30H16O8
MFCD00016683
hypericin, hypericum red, hipericina, hypericine, hyperizin, cyclosan, 1,3,4,6,8,13-hexahydroxy-10,11-dimethylphenanthro 1,10,9,8-opqra perylene-7,14-dione, cyclo-werol, hypericum extract, vimrxyn
CHEBI:5835
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P501c

H302-H317

EINECSNumber : 208-941-0

RTECSNumber : SF8472000

TSCA : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only