Thermo Scientific™ HyperREZ™ XP Carbohydrate Ca2+ HPLC Columns
Manufactured with a proprietary process and subjected to stringent quality control procedures, these columns are used for the appropriate counter-ion to meet application requirements.
Supplier:  Thermo Scientific™ 69208057726
 View more versions of this product
Catalog No. 69-208-057726
Explore available promotions
Description
Description
Manufactured with a proprietary process and subjected to stringent quality control procedures, Thermo Scientific™ HyperREZ™ XP Carbohydrate Ca2+ LC Columns are used for the appropriate counter-ion to meet application requirements. Each column is individually tested to ensure column-to-column reproducibility.
Polymer-based columns for carbohydrate analysis offer benefits to the chromatographer including temperature and pH stability, as well as enhanced solvent choices.
  • Designed for the determination of carbohydrates, saccharides, organic acids and alcohols
  • Efficient and reproducible monodisperse particles
  • Stable for long column lifetimes even at low pH and high temperatures
Specifications
Specifications
Guard Column
LC/MS
725 psi (50 bar)
1 to 11
L19
Spherical, Microporous Polymer
Ligand Exchange
7.7 mm
50 mm
8 μm
90°C
Ca2+ Counter Ion
Carbohydrate Analysis
Guard Column
Videos
SDS
Documents
Documents
Promotions
Promotions
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.