Thermo Scientific™ HyperSep™ Dispersive SPE Clean-up Products (QuEChERS)
Save time and achieve more accurate pesticide determinations using these pre-prepared dispersive SPE products for QuEChERS clean-up.
$214.63 - $890.79
Products 4
Catalog Number Mfr. No. Description For Use With (Application) Volume (Metric) Price Quantity  
Catalog Number Mfr. No. Description For Use With (Application) Volume (Metric) Price Quantity  
60-105-213
View Documents Promotion Details
Thermo Scientific™
60105213
750 mg anhydrous magnesium sulfate, 250 mg PSA, 250 mg C18, 250 mg GCB QuEChERS Method 15 mL
Pack of 50 Each for $890.79
03-150-629
View Documents
Thermo Scientific™
60105208
500 mg PSA on Bottom, 250 mg GCB on Top, Separated by a Frit Schenck Method 6 mL Centrifuge Tubes
Pack of 30 Each for $566.80
03-151-608
View Documents Promotion Details
Thermo Scientific™
60105307
400 mg PSA on Bottom, 200 mg GCB on Top, Separated by a Frit Schenck Method 6 mL Centrifuge Tubes
Pack of 10 Each for $214.63
60-105-232
View Documents Promotion Details
Thermo Scientific™
60105232
150 mg MgSO4, 50 mg PSA, 50 mg Chlorofiltr QuEChERS Method 2 mL Centrifuge Tubes
Pack of 100 Each for $733.32
Description
Description
QuEChERS Dispersive SPE methods offer a convenient and effective approach for extraction and clean-up of pesticide residues in food and other complex matrices. The robust procedure offers a number of compelling advantages: high recoveries, accurate results, high sample throughput, low solvent and glassware usage, reduced labor and bench space and lower reagent costs.
Thermo Scientific™ HyperSep Dispersive SPE Clean-Up Products are available in a range of pre-prepared sorbent combinations, each containing the proper sorbents for optimum clean-up of analytes extracted using QuEChERS methods.
HyperSep Dispersive SPE Clean-Up Products contain PSA (primary/secondary amine) for the removal of organic acids and polar pigments among other compounds. Some products couple the PSA with endcapped C18 for the removal of most lipids and sterols, or graphitized carbon black for the removal of sterols and pigments such as chlorophyll. A variety of tube formats and bed weights are available to accommodate large and small sample sizes.
All HyperSep Dispersive SPE products are pre-prepared and optimized for the method at hand, so you will:
  • Save valuable preparation time
  • Achieve more consistent analytical results due to consistency in products and rigorous quality control
  • Get cleaner extracts and lower peak counts as a result of using highest quality, pre-prepared products
QuEChERS Technique
Developed by the USDA Eastern Regional Research Center in 2003, the QuEChERS (Quick, Easy, Cheap, Effective, Rugged and Safe) technique is increasingly becoming the technique of choice for extraction and clean-up of pesticide residues.
The QuEChERS method is a two-step process: extraction followed by clean-up. There are four variations of the QuEChERS method currently in use:
  • The original QuEChERS method uses sodium chloride to enhance extraction
  • Dispersive AOAC 2007.01 uses sodium acetate as a buffer, replacing sodium chloride
  • The Dual-Phase variation introduces PSA and GCB to remove high levels of chlorophyll and plant sterols in the final extract without the loss of planar pesticides (polar aromatics) using an acetone:toluene solvent mix (3:1)
  • European Version is similar to the AOAC method, except the extraction uses sodium chloride, sodium citrate dihydrate and disodium citrate sesquihydrate instead of sodium acetate
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.