Thermo Scientific™ Hypersil GOLD™ C18 Selectivity HPLC Columns
Use these endcapped, ultrapure, silica-based columns to generate exceptional peak shape and resolution for HPLC and LC/MS analyses using generic gradients
$906.69 - $6194.64
Products 46
Catalog Number Mfr. No. Length (Metric) Diameter (Metric) Particle Size Price Quantity  
Catalog Number Mfr. No. Length (Metric) Diameter (Metric) Particle Size Price Quantity  
25003032130
View Documents
Thermo Scientific™
25003032130
30 mm 2.1 mm 3 μm
Each for $1,567.28
25-003-033030
View Documents
Thermo Scientific™
25003033030
30 mm 3 mm 3 μm
Each for $2,024.04
25-003-034630
View Documents
Thermo Scientific™
25003034630
30 mm 4.6 mm 3 μm
Each for $1,618.38
25003052130
View Documents
Thermo Scientific™
25003052130
50 mm 2.1 mm 3 μm
Each for $1,581.06
03-171-269
View Documents
Thermo Scientific™
25003053030
50 mm 3 mm 3 μm
Each for $1,581.06
25-003-054030
View Documents
Thermo Scientific™
25003054030
50 mm 4 mm 3 μm
Each for $2,238.81
25003054630
View Documents
Thermo Scientific™
25003054630
50 mm 4.6 mm 3 μm
Each for $1,618.38
25003101030
View Documents
Thermo Scientific™
25003101030
100 mm 1 mm 3 μm
Each for $1,832.83
25003102130
View Documents
Thermo Scientific™
25003102130
100 mm 2.1 mm 3 μm
Each for $1,724.59
25003103030
View Documents
Thermo Scientific™
25003103030
100 mm 3 mm 3 μm
Each for $1,724.59
25003104030
View Documents
Thermo Scientific™
25003104030
100 mm 4 mm 3 μm
Each for $1,644.00
25003104630
View Documents
Thermo Scientific™
25003104630
100 mm 4.6 mm 3 μm
Each for $1,764.00
25-003-151030
View Documents
Thermo Scientific™
25003151030
150 mm 1 mm 3 μm
Each for $2,464.62
25003152130
View Documents
Thermo Scientific™
25003152130
150 mm 2.1 mm 3 μm
Each for $1,983.02
25003153030
View Documents
Thermo Scientific™
25003153030
150 mm 3 mm 3 μm
Each for $1,953.88
25003154030
View Documents
Thermo Scientific™
25003154030
150 mm 4 mm 3 μm
Each for $2,020.08
25003154630
View Documents
Thermo Scientific™
25003154630
150 mm 4.6 mm 3 μm
Each for $1,861.05
25003014001
View Documents
Thermo Scientific™
25003014001
10 mm 4 mm 3 μm
Pack of 4 Each for $991.53
03170557
View Documents
Thermo Scientific™
25002012135
10 mm 2.1 mm 1.9 μm
Pack of 3 Each for $1,755.30
25-002-031030
View Documents
Thermo Scientific™
25002031030
30 mm 1 mm 1.9 μm
Each for $1,941.45
25002032130
View Documents
Thermo Scientific™
25002032130
30 mm 2.1 mm 1.9 μm
Each for $1,716.91
25002051030
View Documents
Thermo Scientific™
25002051030
50 mm 1 mm 1.9 μm
Each for $1,881.59
25002052130
View Documents
Thermo Scientific™
25002052130
50 mm 2.1 mm 1.9 μm
Each for $1,873.38
25002053030
View Documents
Thermo Scientific™
25002053030
50 mm 3 mm 1.9 μm
Each for $1,751.94
03170550
View Documents
Thermo Scientific™
25002054630
50 mm 4.6 mm 1.9 μm
Each for $1,889.19
25002101030
View Documents
Thermo Scientific™
25002101030
100 mm 1 mm 1.9 μm
Each for $1,804.48
25002102130
View Documents
Thermo Scientific™
25002102130
100 mm 2.1 mm 1.9 μm
Each for $1,873.38
25002103030
View Documents
Thermo Scientific™
25002103030
100 mm 3 mm 1.9 μm
Each for $1,804.48
03-260-107
View Documents
Thermo Scientific™
25002152130
150 mm 2.1 mm 1.9 μm
Each for $2,132.03
25-002-202130
View Documents
Thermo Scientific™
25002202130
200 mm 2.1 mm 1.9 μm
Each for $1,965.06
25003012101
View Documents
Thermo Scientific™
25003012101
10 mm 2.1 mm 3 μm
Pack of 4 Each for $916.39
25003013001
View Documents
Thermo Scientific™
25003013001
10 mm 3 mm 3 μm
Pack of 4 Each for $916.39
25005012101
View Documents
Thermo Scientific™
25005012101
10 mm 2.1 mm 5 μm
Pack of 4 Each for $906.69
25005014001
View Documents
Thermo Scientific™
25005014001
10 mm 4 mm 5 μm
Pack of 4 Each for $906.69
25005052130
View Documents
Thermo Scientific™
25005052130
50 mm 2.1 mm 5 μm
Each for $1,437.48
25005053030
View Documents
Thermo Scientific™
25005053030
50 mm 3 mm 5 μm
Each for $1,531.94
25-005-103030
View Documents
Thermo Scientific™
25005103030
100 mm 3 mm 5 μm
Each for $1,906.89
25005104630
View Documents
Thermo Scientific™
25005104630
100 mm 4.6 mm 5 μm
Each for $1,774.86
25005152130
View Documents
Thermo Scientific™
25005152130
150 mm 2.1 mm 5 μm
Each for $1,609.45
25005153030
View Documents
Thermo Scientific™
25005153030
150 mm 3 mm 5 μm
Each for $1,609.45
25005154630
View Documents
Thermo Scientific™
25005154630
150 mm 4.6 mm 5 μm
Each for $1,855.17
25-005-252130
View Documents
Thermo Scientific™
25005252130
250 mm 2.1 mm 5 μm
Each for $2,408.04
25005254630
View Documents
Thermo Scientific™
25005254630
250 mm 4.6 mm 5 μm
Each for $1,743.52
25005259070
View Documents
Thermo Scientific™
25005259070
250 mm 10 mm 5 μm
Each for $6,194.64
03-171-256
View Documents
Thermo Scientific™
25003254630
250 mm 4.6 mm 3 μm
Each for $2,132.30
25-002-104630
View Documents
Thermo Scientific™
25002104630
100 mm 4.6 mm 2 μm
Each for $1,629.34
Description
Description
Achieve exceptional peak shape and resolution for your HPLC and LC/MS applications with Thermo Scientific™ Hypersil GOLD™ HPLC columns. These endcapped, ultrapure, silica-based columns deliver significant reduction in peak tailing using generic gradients with C18 selectivity. With their excellent resolution, efficiency, and sensitivity, Hypersil GOLD columns give you confidence in the accuracy and quality of your analytical data.
Novel Bonding
  • Proprietary derivatization and endcapping
  • C18 (USP L1) selectivity
pH Stability
  • Robust performance at high pH
  • Excellent column stability at low pH
Flexible Formats
  • Javelin HTS columns for high-throughput screening
  • Semi-preparative columns with 12 μm particles available
About the Hypersil GOLD Family of HPLC Columns
Meet the challenges of modern chromatography with state-of-the-art columns that provide outstanding performance and consistent separations. Twelve different column chemistries are available so you can optimize separations and maximize productivity. Get outstanding peak shapes in reversed phase, ion exchange, HILIC, or normal phase chromatography.
  • Excellent peak symmetry
  • Narrow peaks for outstanding efficiency
  • Enhanced sensitivity and improved resolution
  • Exceptional reproducibility for reliable chromatography, column after column
  • Solutions for high-throughput screening
  • 1.9, 3, and 5 μm particle sizes for all your separation needs
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.