missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hypoxanthine, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1148106

 View more versions of this product

Catalog No. AAA1148106


Explore available promotions

Description

Description

Hypoxanthine, is used as a marker for energy perturbation in hypoxia/ ischemia. It has been also used in DNA studies to investigate the destabilizing effect it has on DNA duplexes containing hypoxanthine as a base, in a gas phase versus a liquid phase. Hypoxanthine has also been used in studies along with 8-oxoguanine nucleotides to investigate their interaction with human DNA pol A (DNA polymerase alpha) and DNA polymerase I from Bacillus stearothermophilus. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hypoxanthine, is used as a marker for energy perturbation in hypoxia/ ischemia. It has been also used in DNA studies to investigate the destabilizing effect it has on DNA duplexes containing hypoxanthine as a base, in a gas phase versus a liquid phase. Hypoxanthine has also been used in studies along with 8-oxoguanine nucleotides to investigate their interaction with human DNA pol å (DNA polymerase alpha) and DNA polymerase I from Bacillus stearothermophilus. It is also used as a pharmaceutical intermediate.

Solubility
It does not mix appreciably with water (1:1400). Small octahedra crystals from water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to Strong oxidizing agents. Stable under recommended storage conditions. Sterilize by filtration through a 0.22 µm filter.
Specifications

Specifications

Hypoxanthine
>300°C
MFCD00005725
5811
hypoxanthine, 6-hydroxypurine, 9h-purin-6-ol, sarkin, 6-oxopurine, sarcine, sarkine, 3h-purin-6-ol, 7h-purin-6-ol, purin-6 1h-one
FDGQSTZJBFJUBT-UHFFFAOYSA-N
3,7-dihydropurin-6-one
790
136.11
68-94-0
C5H4N4O
5 g
14,4869
It does not mix appreciably with water (1:1400). Small octahedra crystals from water.
O=C1N=CNC2=C1NC=N2
136.11
CHEBI:17368
99%

Chemical Identifiers

68-94-0
136.11
FDGQSTZJBFJUBT-UHFFFAOYSA-N
790
3,7-dihydropurin-6-one
C5H4N4O
MFCD00005725
hypoxanthine, 6-hydroxypurine, 9h-purin-6-ol, sarkin, 6-oxopurine, sarcine, sarkine, 3h-purin-6-ol, 7h-purin-6-ol, purin-6 1h-one
CHEBI:17368
O=C1N=CNC2=C1NC=N2
Videos
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 200-697-3

RTECSNumber : UP0791000

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only