missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Silencer™ Negative Control No. 1 siRNA DFS Item

No significant sequence similarity to mouse, rat, or human gene sequences

Supplier:  Invitrogen™ AM4636

 View more versions of this product

Catalog No. AM4636


Explore available promotions

Description

Description

  • Ambion™ Silencer™ Negative Control #1 siRNA has no significant sequence similarity to mouse, rat, or human gene sequences
  • The control has also been tested in cell-based screens and proven to have no significant effect on cell proliferation, viability, or morphology
  • Validated for use in human, mouse, and rat cells
  • HPLC purified, duplexed, and ready to use
  • Negative control siRNAs are essential for determining transfection efficiency and to control for the effects of siRNA delivery
  • In siRNA screening applications, negative control siRNAs are also critical for setting the hit threshold that determines whether an siRNA is considered to have a positive, negative, or neutral effect in a particular assay
  • Store at or below -20°C in a non-frost-free freezer
Specifications

Specifications

The siRNA is provided dried down in a single tube, along with Nuclease-free Water for resuspension, and should be stored at –20°C.
siRNA
Transfection, RNAi
Silencer™, Ambion™
HPLC
Room Temperature
Negative Control
Human, Mouse, Rat
Unconjugated
siRNA
5 x 40 nmol
Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.