missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ IB™ Model Serum Filters
Store serum up to seven days when it is essential to prevent any exchange of ions.
$152.63 - $162.56
Specifications
Material | Polyethylene |
---|---|
Product Type | Serum Filter |
For Use With (Application) | Eliminates fibrin, particulate matter and equipment downtime |
For Use With (Equipment) | Glass or Plastic collection tubes |
Quantity | 100 Pack |
Catalog Number | Mfr. No. | Diameter (English) Outside | Diameter (Metric) Outside | Length (English) | Length (Metric) | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (English) Outside | Diameter (Metric) Outside | Length (English) | Length (Metric) | Price | Quantity | ||
02-681-66
|
N/A | 0.5 in. | 13mm | 4 in. | 100mm |
Pack of 100 Each for $162.56
Case of 40 Pack for $6,502.40
|
|
||
02-681-65
|
N/A | 0.5 in. | 13mm | 2.75 in. | 70mm |
Pack of 100 Each for $153.74
Case of 40 Pack for $6,149.60
|
|
||
02-681-63
|
N/A | 0.6 in. | 16mm | 4 in. | 100mm |
Pack of 100 Each for $152.63
Case of 40 Pack for $6,105.20
|
|
||
Description
Store serum up to seven days when it is essential to prevent any exchange of ions.
- One-way valve permits a physical gap to be created between the serum/plasma and clot, and prevents backflow into the primary collection tube
- Use with glass or plastic collection tubes
- Technique reduces second centrifugation, tube labeling and sample preparation time
- Allows direct sampling into automatic laboratory equipment
- Eliminates fibrin, particulate matter and equipment downtime
Specifications
Polyethylene | |
Eliminates fibrin, particulate matter and equipment downtime | |
100 Pack |
Serum Filter | |
Glass or Plastic collection tubes |