Learn More
Invitrogen™ iBlot™ 2 Transfer Stacks, nitrocellulose, mini
iBlot 2 Transfer Stacks are used to transfer proteins using the iBlot 2 Gel Transfer Device. They are consumable stacks with an integrated nitrocellulose transfer membrane for dry blotting of proteins.
$266.00 - $266.00
Specifications
Format | Transfer Stack |
---|---|
Material | Nitrocellulose |
Pore Size | 0.2 μm |
Content And Storage | Store at room temperature. |
Dimensions (LxW) | 8 x 8 cm |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
IB23002
|
Invitrogen™
IB23002 |
10 Stacks |
Each for $266.00
|
|
|||||
Description
iBlot 2 Transfer Stacks are used to transfer proteins using the iBlot 2 Gel Transfer Device. They are consumable stacks with an integrated nitrocellulose transfer membrane for dry blotting of proteins. Each iBlot 2 Transfer Stack contains a copper-coated electrode and appropriate cathode and anode buffers in the gel matrix to allow fast, reliable transfer of proteins. This mini-size stack (8 x 8 cm) can be used with one mini-size gel.
Nitrocellulose membrane
The nitrocellulose membrane (0.2-μm pore size) in this stack is composed of 100% pure nitrocellulose to provide high-quality transfer. The membrane is compatible with commonly used detection methods such as staining, immunodetection, fluorescence, and radiolabeling. The proteins bind to the membrane due to hydrophobic and electrostatic interactions. The protein-binding capacity is 209 μg/cm2.
Specifications
Transfer Stack | |
0.2 μm | |
8 x 8 cm | |
Less waste, responsibly packaged, extended life |
Nitrocellulose | |
Store at room temperature. | |
iBlot |
For Research Use Only. Not for use in diagnostic procedures.