missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ibuprofen, USP grade, MP Biomedicals™

Ibuprofen is clinically useful as a nonsteroidal anti-inflammatory drug.

Supplier:  MP Biomedicals Inc 0219021610

 View more versions of this product

Catalog No. ICN19021610


Explore available promotions

Description

Description

  • Cyclooxygenase (COX) inhibitor that has greater activity against COX-1 than against COX-2.
  • Also inhibits cyclooxygenase, PGH synthase-1 and PGH synthase-2.
  • Ibuprofen is reported to inhibit the netrophil-dependent edema response of rabbit skin to C5a/PGE23.
Specifications

Specifications

Ibuprofen
75 to 77.5°C
C13H18O2
10 g
Water Solubility: 21 mg/L (at 25°C)
CC(C)CC1=CC=C(C=C1)C(C)C(O)=O
206.29
CHEBI:5855
4.74X10-5 mm Hg at 25°C
Powder
15687-27-1
White
MFCD00010393
ibuprofen, 2-4-isobutylphenyl propanoic acid, motrin, brufen, nurofen, advil, dolgit, liptan, nuprin, anflagen
HEFNNWSXXWATRW-UHFFFAOYNA-N
2-[4-(2-methylpropyl)phenyl]propanoic acid
3672
206.3
USP

Chemical Identifiers

15687-27-1
206.29
HEFNNWSXXWATRW-UHFFFAOYNA-N
3672
2-[4-(2-methylpropyl)phenyl]propanoic acid
C13H18O2
MFCD00010393
ibuprofen, 2-4-isobutylphenyl propanoic acid, motrin, brufen, nurofen, advil, dolgit, liptan, nuprin, anflagen
CHEBI:5855
CC(C)CC1=CC=C(C=C1)C(C)C(O)=O
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 239-784-6

Recommended Storage : Store at Room Temperature (15 to 30°C)

SDS
Documents

Documents

Promotions

Promotions

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.