Dentec Safety IceTred™ HTD - Heel Traction Devices
IceTred™ HTD - Heel Traction Devices - Run or walk on ice or snow with confidence.
Supplier: Dentec Safety DTC10705
Description
- Tough and Durable Design - Made of durable natural rubber that is incredibly tough but maintains its flexibility in extreme temperatures. Incredible traction on ice but also when transitioning to asphalt and concrete.
- Tiny Solutions BIG GRIP - Six strategically positioned tungsten carbide spikes provide excellent ice traction performance while maintaining comfort under foot. Wears longer and holds its ice piercing edge better than steel spikes or screw heads. Becomes sharper with greater wear. Spark resistant. Used on all ICETRED™ Traction Devices. Rubber Traction studs provide excellent traction on snow
- Adjustable and easy to use - Velcro strap allows the HTD to be easily secured to any footwear regardless of size. Unique heel pad shape forms more naturally to any footwear to ensure maximum spike contact.
- Universal Size - Unique rubber formulation provides more stretch to ensure a tight and secure fit to the footwear regardless of size or type
- Not a Spark Hazard - Third party tested not to be a spark hazard in gas groups A – D as per CSA C22.2 No. 213-17 combustibility testing protocol. If you require intrinsic safety protection from static discharge, must use approved Electrostatic Discharging (ESD) footwear or clothing and/or accessories that offer ESD protection with our ICETRED™ traction devices
Specifications
Rubber | |
Universal | |
Safety Sole |