missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hemosure™ iFOB Test Kits
Qualitative, one-step, odorless iFOB test with high sensitivity and specificity
Supplier: Hemosure™ T1CK50
Description
- CLIA Complexity: Waived
- CPT Codes: G0328QW (colorectal cancer screening); 82274QW (detection of blood in feces form)
- Sensitivity: 50ng hHB/mL
- Specificity: over 96%
- Useful aid in the diagnosis and therapy of gastrointestinal disorders
- Patient-friendly for use in physicians' offices, labs, hospitals, clinics, or homes
- As test sample flows up through absorbent device, the labeled antibody-dye conjugate binds to the hemoglobin in specimen, forming an antibody-antigen complex
- Results within five minutes
- Positive results: one band in the control (C) region, one band in the test (T) region
- Negative results: only one band in the control (C) region
- Low false positive results
- Reaction components: citric acid, monoclonal and polyclonal antibody, mouse anti-HB antibodies 1 and 2, and goat anti-mouse IgG antibody
If home kit mailers are not needed, they can be excluded from the order. Please contact your Fisher HealthCare account manager for details.
Order Info
The 10-test kit contains 10 collection bottles and 10 home kit mailers; 30-test kit contains 50 collection bottles and 50 home kit mailers.
Specifications
G0328QW, 82274QW | |
iFOB Test Kit | |
50 Fecal occult Blood test cassettes, 50 collection tubes | |
5 min. |
Waived | |
Diagnosis and therapy of gastrointestinal disorders | |
50 Pack |