missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rat anti-Mouse IgG2a Secondary Antibody, PerCP-eFluor™ 710, eBioscience™, Invitrogen™

Rat Monoclonal Secondary Antibody

$482.00 - $482.00

Specifications

Antigen Mouse IgG2a
Clone m2a-15F8
Applications Flow Cytometry
Classification Monoclonal
Conjugate PerCP-eFluor 710
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501130807
View Documents
Invitrogen
46421082
100 μg
Each for $482.00
 
Description

Description

Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications

Specifications

Mouse IgG2a
Flow Cytometry
PerCP-eFluor 710
Rat
Mouse
Secondary
m2a-15F8
Monoclonal
Liquid
RUO
IgG1 κ
Affinity chromatography
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use Only.