missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S'il vous plaît vous connecter à votre compte en ligne pour afficher votre prix réduits
Carson™ Illuminated Handheld 7x Power 1.5 in. Aspheric LED Lighted Magnifier (SV-70)
The Illuminated Handheld Magnifier from Carson is a 7x Aspheric magnifier. Aspheric lens magnifiers allow for increased magnification and image clarity while greatly reducing edge to edge distortion. Its 1.5 in. crystal-clear acrylic lens allows it to be lightweight enough to use for everyday tasks.
Supplier: Carson™ SV70A
Description
- The illuminated hand-held magnifier is a great low vision aid and assists in reading small print and other low vision needs.
- It features an ultra bright LED light to brighten your viewing area.
- It is powered by 3 AAA batteries (not included).
Spécifications
2.54 cm | |
Acrylic | |
7x |
Magnifier | |
1.5 in. |