Supelco Illumination Unit
The Illumination Unit contains 3 different wavelength for excitation at 254 nm, 366 nm and VIS light when operating the TLC Explorer Documentation System. The TLC Explorer Documentation System is designed to document and evaluate thin-layer chromatographic (TLC) plates.
Supplier: Supelco 1526130001
Description
- The Illumination unit contains 3 different wavelength for excitation at 254 nm, 366 nm and VIS light when operating the TLC Explorer Documentation System.
- The TLC Explorer Documentation System is designed to document and evaluate thin-layer chromatographic (TLC) plates.
- It offers a safe, fast and flexible way to visually inspect and digitally record images of TLC plates after their chromatographic development workflow in the laboratory.
- For detecting different substance classes, the device contains three different LED light sources (white light (VIS), UVA (366 nm) and UVC (254 nm), which illuminate the plates from above in direct-light mode.
- Recorded plate images can be evaluated to generate reports or export the data for further processing with other systems.
Specifications
Illumination Unit |