missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cytiva NAP™ Columns, NAP-10
Use for rapid and efficient purification of DNA and oligonucleotides (>10-mer) in less than 15 minutes by gravity flow. Cytiva Life Sciences™ illustra™ NAP™ Columns are disposable columns prepacked with Sephadex™ G-25 DNA Grade
$283.00 - $638.00
Specifications
For Use With (Application) | Rapid and efficient purification of DNA and Oligonucleotides |
---|---|
Includes | Complete step-by-step protocol identifying the fixed volumes needed for equilibration, washing and sample elution |
Product Type | NAP-10 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
45-000-153
|
Cytiva
17085401 |
20 Pack |
Pack of 20 Each for $283.00
|
|
||||||
45-000-154
|
Cytiva
17085402 |
50 Pack |
Pack of 50 Each for $638.00
|
|
||||||
Description
- Disposable columns prepacked with Sephadex™ G-25 DNA Grade Gel require only gravity to run
- Available in three sizes depending on sample volume: 0.5mL (NAP-5), 1mL (NAP-10) or 2.5mL (NAP-25)
- Useful for small-scale purification, desalting, and buffer exchange
Specifications
Rapid and efficient purification of DNA and Oligonucleotides | |
Complete step-by-step protocol identifying the fixed volumes needed for equilibration, washing and sample elution | |
NAP-10 |