Learn More
Cytiva NAP™ Columns, NAP-5
Use for rapid and efficient purification of DNA and oligonucleotides (>10-mer) in less than 15 minutes by gravity flow. Cytiva Life Sciences™ illustra™ NAP™ Columns are disposable columns prepacked with Sephadex™ G-25 DNA Grade
Supplier: Cytiva 17085301
Description
- Disposable columns prepacked with Sephadex™ G-25 DNA Grade Gel require only gravity to run
- Available in three sizes depending on sample volume: 0.5mL (NAP-5), 1mL (NAP-10) or 2.5mL (NAP-25)
- Useful for small-scale purification, desalting, and buffer exchange
Specifications
| Rapid and efficient purification of DNA and Oligonucleotides | |
| Complete step-by-step protocol identifying the fixed volumes needed for equilibration, washing and sample elution | |
| NAP-5 | |
| 20 Pack |