missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Iloperidone 98.0+%, TCI America™
$31.14 - $456.75
Chemical Identifiers
CAS | 133454-47-4 |
---|---|
Molecular Formula | C24H27FN2O4 |
Molecular Weight (g/mol) | 426.49 |
MDL Number | MFCD00866688 |
InChI Key | XMXHEBAFVSFQEX-UHFFFAOYSA-N |
PubChem CID | 71360 |
ChEBI | CHEBI:65173 |
IUPAC Name | 1-(4-{3-[4-(6-fluoro-1,2-benzoxazol-3-yl)piperidin-1-yl]propoxy}-3-methoxyphenyl)ethan-1-one |
SMILES | COC1=CC(=CC=C1OCCCN1CCC(CC1)C1=NOC2=CC(F)=CC=C12)C(C)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
I092625MG
|
TCI America
I092625MG |
25 mg |
Each for $31.14
|
|
|||||
I0926100MG
|
TCI America
I0926100MG |
100 mg |
Each for $76.13
|
|
|||||
I09261G
|
TCI America
I09261G |
1 g |
Each for $456.75
|
|
|||||
Chemical Identifiers
133454-47-4 | |
426.49 | |
XMXHEBAFVSFQEX-UHFFFAOYSA-N | |
CHEBI:65173 | |
COC1=CC(=CC=C1OCCCN1CCC(CC1)C1=NOC2=CC(F)=CC=C12)C(C)=O |
C24H27FN2O4 | |
MFCD00866688 | |
71360 | |
1-(4-{3-[4-(6-fluoro-1,2-benzoxazol-3-yl)piperidin-1-yl]propoxy}-3-methoxyphenyl)ethan-1-one |
Specifications
133454-47-4 | |
White | |
MFCD00866688 | |
XMXHEBAFVSFQEX-UHFFFAOYSA-N | |
1-(4-{3-[4-(6-fluoro-1,2-benzoxazol-3-yl)piperidin-1-yl]propoxy}-3-methoxyphenyl)ethan-1-one | |
71360 | |
426.49 | |
Crystalline Powder |
124°C | |
C24H27FN2O4 | |
25 mg | |
COC1=CC(=CC=C1OCCCN1CCC(CC1)C1=NOC2=CC(F)=CC=C12)C(C)=O | |
426.49 | |
CHEBI:65173 | |
≥98.0% (HPLC,T) | |
Iloperidone |
Safety and Handling
RTECSNumber : KM5777850
TSCA : No
Recommended Storage : Freezer (-20 °C)