missing translation for 'onlineSavingsMsg'
Learn More

Imidazole-4-acetic acid monohydrochloride, 97%

CAS: 3251-69-2 | C5H7ClN2O2 | 162.57 g/mol

$204.78 - $679.64

Chemical Identifiers

CAS 3251-69-2
Molecular Formula C5H7ClN2O2
Molecular Weight (g/mol) 162.57
MDL Number MFCD00012698
InChI Key MWHLCFYPFGFBQO-UHFFFAOYSA-N
Synonym 4-imidazoleacetic acid hydrochloride, 2-1h-imidazol-5-yl acetic acid hydrochloride, imidazole-4-acetic acid hydrochloride, 2-1h-imidazol-4-yl acetic acid hydrochloride, 4-imidazoleacetic acid hcl, imidazoleacetic acid hydrochloride, unii-et7a74roao, 4-imidazoleaceticacidhydrochloride, i4aa, et7a74roao
PubChem CID 145685
IUPAC Name 2-(1H-imidazol-5-yl)acetic acid;hydrochloride
SMILES Cl.OC(=O)CC1=CN=CN1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2247303
View Documents
Thermo Scientific Chemicals
B2247303
1 g
Each for $204.78
Only null left
 
AAB2247306
View Documents
Thermo Scientific Chemicals
B2247306
5 g
Each for $679.64
Only null left
 
Description

Description

A GABAc antagonistImidazole-4-acetic acid monohydrochloride is used in the preparation of imidazolyl-polyethylenimine modified nanoparticles. It is also used to prepare pyridyl and imidazoyl functionalized carboproteins and potential metal ion chelators. Further, it acts as an imidazole derivative with potential antituberculousis properties. In addition to this, it is used in the generation of acyl guanidine inhibitors of beta-secretas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A GABAc antagonistImidazole-4-acetic acid monohydrochloride is used in the preparation of imidazolyl-polyethylenimine modified nanoparticles. It is also used to prepare pyridyl and imidazoyl functionalized carboproteins and potential metal ion chelators. Further, it acts as an imidazole derivative with potential antituberculousis properties. In addition to this, it is used in the generation of acyl guanidine inhibitors of beta-secretas.

Solubility
Soluble in water.

Notes
Hygroscopic. Store in cool place. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

3251-69-2
162.57
MWHLCFYPFGFBQO-UHFFFAOYSA-N
145685
Cl.OC(=O)CC1=CN=CN1
C5H7ClN2O2
MFCD00012698
4-imidazoleacetic acid hydrochloride, 2-1h-imidazol-5-yl acetic acid hydrochloride, imidazole-4-acetic acid hydrochloride, 2-1h-imidazol-4-yl acetic acid hydrochloride, 4-imidazoleacetic acid hcl, imidazoleacetic acid hydrochloride, unii-et7a74roao, 4-imidazoleaceticacidhydrochloride, i4aa, et7a74roao
2-(1H-imidazol-5-yl)acetic acid;hydrochloride

Specifications

3251-69-2
Odorless
MFCD00012698
3701591
Soluble in water.
Cl.OC(=O)CC1=CN=CN1
162.57
162.58
Imidazole-4-acetic acid monohydrochloride
221°C to 224°C
C5H7ClN2O2
1 g
4-imidazoleacetic acid hydrochloride, 2-1h-imidazol-5-yl acetic acid hydrochloride, imidazole-4-acetic acid hydrochloride, 2-1h-imidazol-4-yl acetic acid hydrochloride, 4-imidazoleacetic acid hcl, imidazoleacetic acid hydrochloride, unii-et7a74roao, 4-imidazoleaceticacidhydrochloride, i4aa, et7a74roao
MWHLCFYPFGFBQO-UHFFFAOYSA-N
2-(1H-imidazol-5-yl)acetic acid;hydrochloride
145685
97%
Videos
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 221-840-6

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only