missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Imidazole-4-sulfonyl chloride, 97%, Thermo Scientific Chemicals

$164.90 - $2173.15

Chemical Identifiers

CAS 58767-51-4
Molecular Formula C3H3ClN2O2S
Molecular Weight (g/mol) 166.579
MDL Number MFCD08687938
InChI Key MXCZCMCHRWGSBX-UHFFFAOYSA-N
Synonym 1h-imidazole-4-sulfonyl chloride, 4-chlorosulfonylimidazole, imidazole-4-sulfonyl chloride, pubchem19106, imidazole-4-sulfonylchloride, imidazole-4-sulphonyl chloride, imidazole-4-sul-fonyl chloride, 1-h-imidazole-4-sulphonyl chloride
PubChem CID 18961164
IUPAC Name 1H-imidazole-5-sulfonyl chloride
SMILES C1=C(NC=N1)S(=O)(=O)Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5464203
View Documents
Thermo Scientific Chemicals
H5464203
1 g
Each for $164.90
 
AAH5464206
View Documents
Thermo Scientific Chemicals
H5464206
5 g
Each for $602.64
 
AAH5464214
View Documents
Thermo Scientific Chemicals
H5464214
25 g
Each for $2,173.15
 
Description

Description

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Slightly soluble in water. 

Notes
Store at room temperature. Keep away from oxidizing agents and acids. Hygroscopic.
Specifications

Chemical Identifiers

58767-51-4
166.579
MXCZCMCHRWGSBX-UHFFFAOYSA-N
18961164
C1=C(NC=N1)S(=O)(=O)Cl
C3H3ClN2O2S
MFCD08687938
1h-imidazole-4-sulfonyl chloride, 4-chlorosulfonylimidazole, imidazole-4-sulfonyl chloride, pubchem19106, imidazole-4-sulfonylchloride, imidazole-4-sulphonyl chloride, imidazole-4-sul-fonyl chloride, 1-h-imidazole-4-sulphonyl chloride
1H-imidazole-5-sulfonyl chloride

Specifications

58767-51-4
MFCD08687938
UN3261
1h-imidazole-4-sulfonyl chloride, 4-chlorosulfonylimidazole, imidazole-4-sulfonyl chloride, pubchem19106, imidazole-4-sulfonylchloride, imidazole-4-sulphonyl chloride, imidazole-4-sul-fonyl chloride, 1-h-imidazole-4-sulphonyl chloride
MXCZCMCHRWGSBX-UHFFFAOYSA-N
1H-imidazole-5-sulfonyl chloride
18961164
97%
C3H3ClN2O2S
1 g
Moisture sensitive
Slightly soluble in water.
C1=C(NC=N1)S(=O)(=O)Cl
166.579
165.96
Imidazole-4-sulfonyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Hazard Class: 8; Packaging Group: III

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only