missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Imidazole (Certified), Fisher Chemical

Supplier:  Fisher Chemical O3196500

Catalog No. O3196-500

Explore available promotions

Description

Description

Specifications

Specifications

Imidazole
86°C
10.5
145°C
Poly Bottle
NHCH:NCH:CH
500 g
imidazole, glyoxaline, imidazol, iminazole, miazole, 1,3-diazole, glyoxalin, imutex, 1,3-diaza-2,4-cyclopentadiene, pyrro b monazole
N1C=CN=C1
68.08
CHEBI:16069
0.003mbar at 20°C
Certified
288-32-4
White-Yellow
255°C
≥99 %
C3H4N2
MFCD00005183
3263
RAXXELZNTBOGNW-UHFFFAOYSA-N
1H-imidazole
795
68.08
≥99%
Solid

Chemical Identifiers

288-32-4
68.08
RAXXELZNTBOGNW-UHFFFAOYSA-N
795
1H-imidazole
C3H4N2
MFCD00005183
imidazole, glyoxaline, imidazol, iminazole, miazole, 1,3-diazole, glyoxalin, imutex, 1,3-diaza-2,4-cyclopentadiene, pyrro b monazole
CHEBI:16069
N1C=CN=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Causes burns by all exposure routes. Harmful if swallowed. Possible risk of harm to the unborn child.
NFPA
Health:3 Flammability:1 Instability:0

Signal Word: DANGER!

DOTInformation : Class 8: Corrosive

SDS Documents

Documents

Promotions

Promotions