Learn More
Thermo Scientific Chemicals Iminodiacetic acid, 98+%
$65.76 - $91.24
Chemical Identifiers
CAS | 142-73-4 |
---|---|
Molecular Formula | C4H7NO4 |
Molecular Weight (g/mol) | 133.1 |
MDL Number | MFCD00004280 |
InChI Key | NBZBKCUXIYYUSX-UHFFFAOYSA-N |
Synonym | iminodiacetic acid, 2,2'-iminodiacetic acid, glycine, n-carboxymethyl, aminodiacetic acid, diglycocoll, diglykokoll, diglycin, iminodiethanoic acid, n-carboxymethyl glycine, 2,2'-azanediyldiacetic acid |
PubChem CID | 8897 |
ChEBI | CHEBI:24786 |
IUPAC Name | 2-(carboxymethylamino)acetic acid |
SMILES | C(C(=O)O)NCC(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC204970250
|
Thermo Scientific Chemicals
204970250 |
25 g |
Each for $65.76
|
|
|||||
AC204975000
|
Thermo Scientific Chemicals
204975000 |
500 g |
Each for $91.24
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
142-73-4 | |
133.1 | |
NBZBKCUXIYYUSX-UHFFFAOYSA-N | |
8897 | |
2-(carboxymethylamino)acetic acid |
C4H7NO4 | |
MFCD00004280 | |
iminodiacetic acid, 2,2'-iminodiacetic acid, glycine, n-carboxymethyl, aminodiacetic acid, diglycocoll, diglykokoll, diglycin, iminodiethanoic acid, n-carboxymethyl glycine, 2,2'-azanediyldiacetic acid | |
CHEBI:24786 | |
C(C(=O)O)NCC(=O)O |
Specifications
142-73-4 | |
White or Yellow | |
98+% | |
C4H7NO4 | |
MFCD00004280 | |
04, 365 | |
iminodiacetic acid, 2,2'-iminodiacetic acid, glycine, n-carboxymethyl, aminodiacetic acid, diglycocoll, diglykokoll, diglycin, iminodiethanoic acid, n-carboxymethyl glycine, 2,2'-azanediyldiacetic acid | |
NBZBKCUXIYYUSX-UHFFFAOYSA-N | |
2-(carboxymethylamino)acetic acid | |
8897 | |
133.1 | |
Crystalline Powder |
243°C | |
Authentic | |
Plastic bottle | |
HN(CH2CO2H)2 | |
25 g | |
15, 4959 | |
Solubility in water: 2.43g/100 mL water (5°C). Other solubilities: 2.9g/100 mL water (20°C),6.3g/100 mL water (40°C),12.8g/100 mL water (60°C),23.0g/100 mL water (80°C),32.5g/100 mL water (100°C) | |
C(C(=O)O)NCC(=O)O | |
133.1 | |
CHEBI:24786 | |
98+% | |
Iminodiacetic acid, 98+% |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 205-555-4
RTECSNumber : AI2975000
TSCA : TSCA
RUO – Research Use Only