Learn More
Imipenem monohydrate, 95%, Thermo Scientific Chemicals
Imipenem monohydrate,Tienam, Primaxin, CAS # 74431-23-5 is a broad-spectrum carbapenem antibiotic that interferes with the activity of penicillin binding proteins along the inner cell wall.
Supplier: Thermo Scientific Chemicals J66749MB
Description
Imipenem monohydrate is a broad spectrum B-lactam antibiotic.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
- Imipenem monohydrate is a beta-lactam derived from the compound thienamycin, produced by Streptomyces cattleya
- Disrupts cell wall integrity, preventing cell division and promoting lysis by interfering with the activity of penicillin binding proteins
- Imipenem is a third-generation cephalosporin that can kill both gram-negative and gram-positive bacteria
Applications
- Suitable for the study of antibiotic resistance
- High-quality, laboratory grade for use in vitro experiments
Specifications
Imipenem Monohydrate | |
Powder | |
193°C to 198°C | |
25 mg | |
C12H19N3O5S | |
567°C | |
Soluble in water at 5mg/ml | |
O.C[C@H](O)[C@@H]1[C@H]2CC(SCCN=CN)=C(N2C1=O)C(O)=O | |
317.36 | |
CHEBI:51799 | |
95% |
Imipenem monohydrate | |
74431-23-5 | |
Plastic Bottle | |
>110°C (230°F) | |
MFCD09753321 | |
Primaxin,Tienam | |
GSOSVVULSKVSLQ-XMCAPODCSA-N | |
(5R,6S)-3-({2-[(aminomethylidene)amino]ethyl}sulfanyl)-6-[(1S)-1-hydroxyethyl]-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid hydrate | |
5282372 | |
317.36 |
Chemical Identifiers
74431-23-5 | |
317.36 | |
GSOSVVULSKVSLQ-XMCAPODCSA-N | |
CHEBI:51799 | |
O.C[C@H](O)[C@@H]1[C@H]2CC(SCCN=CN)=C(N2C1=O)C(O)=O |
C12H19N3O5S | |
MFCD09753321 | |
5282372 | |
(5R,6S)-3-({2-[(aminomethylidene)amino]ethyl}sulfanyl)-6-[(1S)-1-hydroxyethyl]-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid hydrate |
Safety and Handling
EINECSNumber : 264-734-5
RTECSNumber : CL5446516
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only