missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Imipramine hydrochloride, Thermo Scientific Chemicals

A serotonin uptake inhibitor

$126.92 - $418.19

Chemical Identifiers

CAS 113-52-0
Molecular Formula C19H25ClN2
Molecular Weight (g/mol) 316.873
MDL Number MFCD00012669
InChI Key XZZXIYZZBJDEEP-UHFFFAOYSA-N
Synonym imipramine hydrochloride, imipramine hcl, tofranil, chimoreptin, chrytemin, deprinol, feinalmin, imilanyle, persamine, pertofram
PubChem CID 8228
ChEBI CHEBI:5882
IUPAC Name 3-(5,6-dihydrobenzo[b][1]benzazepin-11-yl)-N,N-dimethylpropan-1-amine;hydrochloride
SMILES CN(C)CCCN1C2=CC=CC=C2CCC3=CC=CC=C31.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6372306
View Documents
Thermo Scientific Chemicals
J6372306
5 g
Each for $126.92
 
AAJ6372314
View Documents
Thermo Scientific Chemicals
J6372314
25 g
Each for $418.19
 
Description

Description

Imipramine hydrochloride is used as a serotonin uptake inhibitor. It mainly used in the treatment of major depression and enuresis (inability to control urination). It has also been evaluated for use in panic disorder.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Imipramine hydrochloride is used as a serotonin uptake inhibitor. It mainly used in the treatment of major depression and enuresis (inability to control urination). It has also been evaluated for use in panic disorder.

Solubility
Soluble in water

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

113-52-0
316.873
XZZXIYZZBJDEEP-UHFFFAOYSA-N
8228
3-(5,6-dihydrobenzo[b][1]benzazepin-11-yl)-N,N-dimethylpropan-1-amine;hydrochloride
C19H25ClN2
MFCD00012669
imipramine hydrochloride, imipramine hcl, tofranil, chimoreptin, chrytemin, deprinol, feinalmin, imilanyle, persamine, pertofram
CHEBI:5882
CN(C)CCCN1C2=CC=CC=C2CCC3=CC=CC=C31.Cl

Specifications

113-52-0
White
MFCD00012669
14,4920
Soluble in water
CN(C)CCCN1C2=CC=CC=C2CCC3=CC=CC=C31.Cl
316.873
CHEBI:5882
Crystalline Powder
174°C to 175°C
C19H25ClN2
5 g
imipramine hydrochloride, imipramine hcl, tofranil, chimoreptin, chrytemin, deprinol, feinalmin, imilanyle, persamine, pertofram
XZZXIYZZBJDEEP-UHFFFAOYSA-N
3-(5,6-dihydrobenzo[b][1]benzazepin-11-yl)-N,N-dimethylpropan-1-amine;hydrochloride
8228
316.87
Imipramine hydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 204-030-7

RTECSNumber : HO1925000

TSCA : Yes

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only