missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MilliporeSigma™ Immobilon™-E Transfer Membrane
Immobilon™-E provides the a PVDF blotting membrane that wets in transfer buffer or water and eliminates the need for the alcohol pre-wet step.
$59.40 - $775.00
Specifications
Description | Easily wetted with water |
---|---|
Color | White |
Code | IEVH |
Surface | Plain |
Material | PVDF |
Catalog Number | Mfr. No. | Dimensions (LxW) | Quantity | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (LxW) | Quantity | Price | Quantity | ||||
IEVH00005
|
MilliporeSigma™
IEVH00005 |
1.875 m x 26.5 cm | 1 Roll |
Each for $373.00
|
|
||||
IEVH85R
|
MilliporeSigma™
IEVH85R |
10 m x 8.5 cm | 1 Roll |
Each for $632.00
|
|
||||
IEVH07804
|
MilliporeSigma™
IEVH07804 |
7 x 8.4 cm | 4 Sheets |
Each for $59.40
|
|
||||
IEVH07850
|
MilliporeSigma™
IEVH07850 |
7 x 8.4 cm | 50 Sheets |
Each for $775.00
|
|
||||
IEVH08100
|
MilliporeSigma™
IEVH08100 |
8 x 10 cm | 10 Sheets |
Each for $244.00
|
|
||||
IEVH09120
|
MilliporeSigma™
IEVH09120 |
9 x 12 cm | 10 Sheets |
Each for $244.00
|
|
||||
IEVH10100
|
MilliporeSigma™
IEVH10100 |
10 x 10 cm | 10 Sheets |
Each for $263.00
|
|
||||
Description
Features:
- 0.45 μm pore size is ideal for most Western blotting applications with chemiluminescence or chromogenic detection
- Saves time and reduces waste by eliminating the alcohol pre-wet step
- Higher protein retention delivers stronger signal than most nitrocellulose membranes
Specifications
Easily wetted with water | |
IEVH | |
PVDF |
White | |
Plain | |
0.45 μm |