Immunostics Hema-Screen™ Fecal Occult Blood Test Kit
For detecting occult blood in stool
Supplier:  Immunostics HS34C
Catalog No. 23-038-050
Explore available promotions
Includes:
  • 34 hema-screen triple slides with positive and negative on-slide controls
  • 2 x 10ml bottles hema-screen™ FOBT developer
  • 102 Wood applicators sticks
  • 1 Product insert
Description
Description
  • CLIA WaivedGÇï
  • Detection level: 0.6mg Hb/gm of feces
  • Routine hospital testing
  • Mass screening programs for colorectal cancer
  • Testing of postoperative patients and newborn infants
Specifications
Specifications
Fecal Occult Blood Test Kit
  • 34 hema-screen triple slides with positive and negative on-slide controls
  • 2 x 10ml bottles
  • hema-screen
  • FOBT developer
  • 102 Wood applicators sticks
  • 1 Product insert