Impacto™ Air Tennis Elbow Support
Supplier:  Impacto™ AIRELBOW
Catalog No. 19-084746
Explore available promotions
Includes: Velcro™ closure.
Description
Description
Designed to support the upper forearm during activities involving repeated hand motion, especially those requiring a strong grip, torquing of the arm, and impact. Such activities include tennis and hand tool use. Inflatable chambered air system provides concentrated compression to a large area, including the extensor muscles of the forearm. Separate air chambers provide excellent conformation to the arm, dampening vibration and shock to the lateral epicondyle tendon.
Specifications
Specifications
Hook and loop
Universal
Nylon
Elbow Support
To support the upper forearm during activities involving repeated hand motion, especially those requiring a strong grip, torquing of the arm and impact