Impacto™ Ergomate Tracmates Anti-Slip Safety Soles
Impacto™ Ergomates Tracmates are used on any slippery and wet surfaces where there is risk of injury due to falling
$186.06 - $186.06
Specifications
Color Black, Green
For Use With (Application) Stripping wax from floor, washing/scrubbing floors, slick kitchen ceramic tiles, slippery concrete, food processing areas, wet surfaces
Material EVA
Toe Material Plain Toe
Gender Unisex
View More Specs
Products 1
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19-500-182
View Documents
Impacto™
G88602
X-Large
Case of 12 Each for $186.06
Description
Description
Impacto™ Ergomate Tracmates Strap-On Soles
  • Reusable design, ideal for any person who is stripping, washing, or scrubbing floors; food processing, working on slippery surfaces; custodial and janitorial workers, hospitals, kitchens, and health care facilities
  • Not recommended for use on dry floors
  • Gain greater traction when: stripping wax floors, washing and scrubbing surfaces, on slick ceramic tiles, working on slippery concrete, walking in food processing areas, working on wet surfaces
Specifications
Specifications
Black, Green
EVA
Unisex
Stripping wax from floor, washing/scrubbing floors, slick kitchen ceramic tiles, slippery concrete, food processing areas, wet surfaces
Plain Toe
Safety Soles
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'