Impacto™ IMPACTOE Steel Toe Cap Overshoes
Impacto™ Safety Toe Cap protects the toe area and is ideal for any working environment that carries a risk of accidental toe damage or has a steel toe cap requirement.
Supplier:  Impacto™ IMPACTOEXXS
 View more versions of this product
Catalog No. 19-400-146
Explore available promotions
Description
Description
Impacto™ IMPACTOE Steel toe cap Overshoes
  • Designed to fit tightly over existing shoe
  • Made of vulcanized rubber
  • Flat anti-slip under sole for greasy and oily environment
  • Waterproof - will not harden, crack
  • Acid, oil and animal fat resistant
  • Pull-on-tab and wide shank for true slip-on entry to provide stretch
  • Recommended for use in construction and heavy work environments
  • Transferable from person to person without concern for hygiene or size restraints
  • Color-coded toe for easy identification of sizes
Specifications
Specifications
CE
Vulcanized Rubber
Steel
Yes
Black
Men: 5.5 to 6, Women: 7 to 7.5
Unisex
IMPACTOE safety toe cap