Fischer Technical Company™ In-line Vacuum Gauge with Hose Barbs DFS Item
With two 3/8 in. barbs, this gauge allows vacuum reading in the middle of your tubing setup.
Supplier:  Fischer Technical Company™ HVG002
Catalog No. S012381
Explore available promotions
Alert:
Prop 65: Cancer and Reproductive Harm.
Description
Description
  • In-line 0 to 30 in. Hg Bourdon Tube Type Vacuum Gauge with two 3/8 in. hose barbs
  • For best results, use hose clamps to secure the vacuum tubing to the hose barbs
Specifications
Specifications
±3% Full-scale
Vacuum/Pressure Gauge
0 to 30 mmHg
Analog