missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Indene 98.0+%, TCI America™
Supplier: TCI America I035425ML
Specifications
Indene | |
Yellow | |
C9H8 | |
25 mL | |
indene, indonaphthene, inden, unii-67h8y6lb8a, indenyl radical, 1h-inden, pubchem20728, indene, reag, acmc-209ruy | |
C1C=CC2=CC=CC=C21 | |
116.163 | |
CHEBI:41921 | |
≥98.0% (GC) |
95-13-6 | |
180°C | |
MFCD00003777 | |
3295 | |
YBYIRNPNPLQARY-UHFFFAOYSA-N | |
1H-indene | |
7219 | |
116.16 | |
Liquid |
Chemical Identifiers
95-13-6 | |
116.163 | |
YBYIRNPNPLQARY-UHFFFAOYSA-N | |
7219 | |
1H-indene |
C9H8 | |
MFCD00003777 | |
indene, indonaphthene, inden, unii-67h8y6lb8a, indenyl radical, 1h-inden, pubchem20728, indene, reag, acmc-209ruy | |
CHEBI:41921 | |
C1C=CC2=CC=CC=C21 |
Safety and Handling
EINECSNumber : (4)-0580
RTECSNumber : NK8225000
TSCA : Yes
Recommended Storage : Freezer