missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Indigo Carmine, Reagent, ACS, Spectrum™ Chemical
I3281, 860-22-0, C16H8N2O8S2Na2
$446.27 - $4,825.83
Chemical Identifiers
| CAS | 860-22-0 |
|---|---|
| Molecular Formula | C16H8N2Na2O8S2 |
| Molecular Weight (g/mol) | 466.35 |
| MDL Number | MFCD00005723 |
| InChI Key | KHLVKKOJDHCJMG-QDBORUFSSA-L |
| IUPAC Name | disodium (E)-3,3'-dioxo-1H,1'H,3H,3'H-[2,2'-biindolylidene]-5,5'-disulfonate |
| SMILES | [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2N\C(C(=O)C2=C1)=C1\NC2=CC=C(C=C2C1=O)S([O-])(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18603437
|
Spectrum Chemical Mfg Cor
I328125GM |
25 g | Amber Glass Bottle |
N/A
|
|
||||
|
18603438
|
Spectrum Chemical Mfg Cor
I3281100GM |
100 g | Amber Glass Bottle |
N/A
|
|
||||
|
18603439
|
Spectrum Chemical Mfg Cor
I32811KGBL |
1 kg | Amber Glass Bottle |
N/A
|
|
||||
Description
Spectrum™ Chemical Indigo Carmine, Reagent, ACS , also known as 5,5′-indigodisulfonic acid sodium salt, is a pH indicator. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.Chemical Identifiers
| 860-22-0 | |
| 466.35 | |
| KHLVKKOJDHCJMG-QDBORUFSSA-L | |
| [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2N\C(C(=O)C2=C1)=C1\NC2=CC=C(C=C2C1=O)S([O-])(=O)=O |
| C16H8N2Na2O8S2 | |
| MFCD00005723 | |
| disodium (E)-3,3'-dioxo-1H,1'H,3H,3'H-[2,2'-biindolylidene]-5,5'-disulfonate |
Specifications
| 860-22-0 | |
| C16H8N2Na2O8S2 | |
| KHLVKKOJDHCJMG-QDBORUFSSA-L | |
| disodium (E)-3,3'-dioxo-1H,1'H,3H,3'H-[2,2'-biindolylidene]-5,5'-disulfonate | |
| ACS | |
| Dark Violet/Blue | |
| Indigo Carmine |
| 100% | |
| MFCD00005723 | |
| [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2N\C(C(=O)C2=C1)=C1\NC2=CC=C(C=C2C1=O)S([O-])(=O)=O | |
| 466.35 | |
| Amber Glass Bottle | |
| 25 g |