missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Troemner™ Individual Analytical Precision Weights, Class 1
Low magnetic susceptibility, uniform consistency and high resistance to corrosion
Supplier: Troemner™ 30391403
Description
- Used in extremely stable environments
- 1mg to 500mg weights are one-piece construction with no adjusting cavity with one end turned up for easy handling
- 1g to 5kg are two-piece construction with handling knob
- Surfaces are polished to a perfect, mirror like finish
- Can be used as a reference standard in calibrating other weights
- Appropriate for calibrating high-precision analytical balances with a readability as low as 0.1 mg to 0.01mg
Certifications
A NIST/NVLAP™ weight calibration certificate or traceable certificate may be purchased to meet all of your ISO™, GLP, GMP, FDA, or nuclear requirements
Specifications
One Piece Metric Analytical Sheet Weight | |
Stainless Steel | |
7.95g/cm3 at 20°C | |
Flat with one end turned up for easy handling | |
Each |
ASTM Class 1 | |
300 mg | |
300 mg | |
Polycarbonate case | |
1 |