Troemner™ Individual Analytical Precision Weights, Class 2
Designed for your most demanding weighing applications
$230.29 - $3585.12
Specifications
Classification ASTM Class 2
Includes Polycarbonate case
Quantity Each
Packaging Quantity 1
View More Specs
Products 17
Catalog Number Mfr. No. Mass Price Quantity  
Catalog Number Mfr. No. Mass Price Quantity  
02-217-2A
View Documents
N/A 1 g
Each for $271.59
02-217-3F
View Documents
N/A 500 mg
Each for $243.31
02-214-023
View Documents
Troemner™
80858368
300 mg
Each for $230.29
02-217-4A
View Documents
N/A 1 mg
Each for $243.31
02-217-3A
View Documents
N/A 10 mg
Each for $243.31
02-217-2G
View Documents
N/A 100 g
Each for $358.70
02-215-78
View Documents
Troemner™
80858165
200 g
Each for $533.15
02-214-027
View Documents
Troemner™
80858111
2 kg
Each for $2,361.91
02-217-4C
View Documents
N/A 5 mg
Each for $235.20
02-217-3E
View Documents
N/A 200 mg
Each for $243.31
02-214-024
View Documents
Troemner™
80858272
3 g
Each for $262.00
02-217-2F
View Documents
N/A 50 g
Each for $343.43
022172D
View Documents
Troemner™
80858240
10 g
Each for $260.21
02-214-026
View Documents
Troemner™
80858124
1 kg
Each for $1,501.21
02-217-3D
View Documents
N/A 100 mg
Each for $235.20
02214028
View Documents
Troemner™
80858084
5 kg
Each for $3,585.12
02214025
View Documents
Troemner™
80858138
500 g
Each for $702.79
Description
Description
Low magnetic susceptibility, uniform consistency and high resistance to corrosion. Analytical Precision Weights meet or exceed specifications as defined in the ANSI/ASTM E617 standard.
  • Used in extremely stable environments
  • 1mg to 500mg weights are one-piece construction with no adjusting cavity with one end turned up for easy handling
  • 1g to 5kg are two-piece construction with handling knob
  • Surfaces are polished to a perfect, mirror like finish
  • Appropriate for calibrating high-precision top loading balances with a readability as low as 0.01 g to 0.001g
  • Weights 1mg through 2kg are supplied in a high quality polycarbonate case
Specifications
Specifications
ASTM Class 2
Polycarbonate case
Each
1
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'